missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA1984 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | KIAA1984 |
---|---|
Concentration | 0.26 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16810535
|
Proteintech
20945-1-AP-20UL |
20 μL |
140.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16800535
|
Proteintech
20945-1-AP-150UL |
150 μL |
399.00 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCDC183 is a protein coding gene.Specifications
KIAA1984 | |
Immunofluorescence, Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q5T5S1 | |
84960 | |
KIAA1984 Fusion Protein Ag15053 | |
Primary | |
-20°C | |
CCDC183 |
0.26 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
KIAA1984, PARF, RP11 216L13.9 | |
CCDC183 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |