Learn More
DWK Life Sciences Kimble™ KIMAX™ Reusable Tubes with PTFE-Faced Rubber-Lined Screw Cap
Click to view available options
Diameter (Metric):
13 mm
16 mm
20 mm
25 mm
Length (Metric):
100 mm
125 mm
150 mm
200 mm
Capacity (Metric):
8 mL
12 mL
16 mL
20 mL
25 mL
30 mL
50 mL
70 mL
Thread Size:
13-415
15-415
18-415
24-410
Unit Size:
Pack of 144
Pack of 192
Pack of 288
Description
Black phenolic cap with PTFE-faced rubber liner
- White marking spot
- Black phenolic cap
Specifications
Specifications
| Certifications/Compliance | ASTM E982, Type VI, Class B |
| Closure Color | Black |
| Closure Material | Phenolic |
| Capacity (Metric) | 20 mL |
| Length (Metric) | 150 mm |
| Material | Borosilicate Glass |
| Label Type | Marking Spot |
| Liner Material | PTFE, Rubber |
| Thread Size | 15-415 |
| Diameter (Metric) | 16 mm |
| Show More |