Learn More
DWK Life Sciences Kimble™ AmberScrew Thread Vials without Closures
Manufactured from amber 33 expansion borosilicate glass
Brand: DWK Life Sciences 609121738
This item is not returnable.
View return policy
Description
- Amber vials offer protection against harmful light and UV rays for light sensitive products.
Specifications
Amber | |
Borosilicate Glass | |
Screw | |
Screw Cap | |
USP Type I |
17 mm | |
4 mL | |
1 dr. | |
38 mm | |
15-425 |