Learn More
DWK Life Sciences Kimble™ Clear Screw Thread Sample Vials without Closures
Packaged in corrugated trays with partitions
Brand:  DWK Life Sciences 608121760
 View more versions of this product
Product Code. 11872983
  • 250.00 EUR / Pack of 200
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Description
  • Manufactured from clear 33 expansion borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Borosilicate Glass
Screw
60 mm
15-425
17 mm
8 mL
2.16 dr.
ASTM Type I, Class A