Learn More
DWK Life Sciences Kimble™ KIMAX™ Serialized/Certified Cylindrical Microflasks
Calibrated To Contain
Brand:  DWK Life Sciences 28017A-10
 View more versions of this product
Product Code. 11848492
  • 703.00 EUR / Pack of 6
Estimated Shipment: 01-04-2025
to see stock.
Explore more special offers
Description
Calibrated To Contain
  • Permanently marked with individual serial number
  • Individually calibrated and certified against equipment whose calibration is traceable to NIST™
  • Cylindrical body for better mixing and draining
  • Easier to withdraw samples using pipets
  • Wide base provides stability
  • Marking spot
  • Ground glass stopper
  • 1 and 2mL sizes come with standard taper 8 stopper; 10mL size with standard taper 9
stopper
Compliance
Designed from requirements of ASTM™ E 237, Class A serialized.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 mL