Learn More
DWK Life Sciences Kimble™ Kontes™ Dounce Tissue Grinders
Click to view available options
Capacity:
0.01 oz., 0.5mL
0.03 oz., 1mL
0.06 oz., 2mL
0.23 oz., 7mL
0.5 oz., 15mL
1.35 oz., 40mL
3.3 oz., 100mL
Clearance (English):
0.0005 to 0.0025 in.
0.0005 to 0.0055 in.
0.0008 to 0.0022 in.
0.001 to 0.003 in.
Length (English):
1.4 in.
2.3 in.
3.4 in.
4.9 in.
6.1 in.
8.4 in.
9.1 in.
Unit Size:
Each
Description
Designed for cellular work in which the nucleus remains intact after homogenization
All-glass construction
Order Info
Items are sold one per case. Supplied with two pestles: large (for initial sample reduction) and small (to form the final homogenate)
Specifications
Specifications
| Clearance (English) | 0.0008 to 0.0022 in. |
| Clearance (Metric) | 0.071 to 0.11mm |
| Capacity (English) | 0.23 oz. |
| Capacity (Metric) | 7 mL |
| Material | Glass |
| Product Type | Dounce Tissue Grinder |
| For Use With (Application) | Designed for cellular work in which the nucleus remains intact after homogenization |
| Length (English) | 4.9 in. |
| Length (Metric) | 125mm |
| Capacity | 0.23 oz., 7mL |