Learn More
DWK Life Sciences Kimble™ Kontes™ FlexColumn Economy Columns Flow Adapters
Convert standard FlexColumn™ columns to adjustable bed volume columns
Brand:  DWK Life Sciences 420415-1000
 View more versions of this product
Product Code. 11798232
Description
Description
Includes
  • PTFE eluent cannula
  • Polyacetal body
  • Viton™ O-ring seal
  • One replacement 20μm polyethylene bed support
Order Info
Additional bed supports can be ordered in packs of 10 (see K420415-1001, -1501, -2501).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Concentration and pH gradients, reverse flow and gravity flow separation
Flow Control Adapter
Polyacetal
0.062 in. O.D. FEP PTFE tubing, Viton™ FKM O-ring seal, replacement 20um porosity HDPE bed support
10 mm I.D. FlexColumn
140 mm
HDPE