Learn More
DWK Life Sciences Kimble™ Kontes™ PTFE-Plugged Columns
Product Code. 11837473
Click to view available options
Length (Metric):
150 mm
200 mm
250 mm
300 mm
350 mm
400 mm
500 mm
700 mm
Diameter (Metric) Inner:
11 mm
15 mm
19 mm
22 mm
25 mm
30 mm
41 mm
49 mm
Column Capacity:
10 mL
20 mL
30 mL
40 mL
80 mL
110 mL
130 mL
140 mL
150 mL
190 mL
240 mL
250 mL
300 mL
650 mL
1300 mL
Includes:
2mm Bore PTFE Stopcock Plug
Fritted Disc, 2mm Bore PTFE Stopcock Plug
Unit Size:
Each
Product Code. 11837473
Brand: DWK Life Sciences 4205300252
Only null left
Add to basket
Add to basket
Manufactured from borosilicate glass 3.3
With PTFE stopcock plug
  • Sealed-in, coarse porosity (40 to 60μm) fritted disc supports packing
  • Use glass wool to support packing in columns without fritted disc
  • PTFE plug controls flowrate
  • Packing not included
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Analytical separation
Diameter (Metric) Inner 30 mm
Type Plain Chromatography Column
Column Capacity 250 mL
Length (Metric) 400 mm
Material Glass
Support Material Glass Wool
Includes 2mm Bore PTFE Stopcock Plug