Learn More
DWK Life Sciences Kimble™ Kontes™ PTFE-Plugged Columns
Click to view available options
Length (Metric):
150 mm
200 mm
250 mm
300 mm
350 mm
400 mm
500 mm
700 mm
Diameter (Metric) Inner:
11 mm
15 mm
19 mm
22 mm
25 mm
30 mm
41 mm
49 mm
Column Capacity:
10 mL
20 mL
30 mL
40 mL
80 mL
110 mL
130 mL
140 mL
150 mL
190 mL
240 mL
250 mL
300 mL
650 mL
1300 mL
Includes:
2mm Bore PTFE Stopcock Plug
Fritted Disc, 2mm Bore PTFE Stopcock Plug
Unit Size:
Each
Description
With PTFE stopcock plug
- Sealed-in, coarse porosity (40 to 60μm) fritted disc supports packing
- Use glass wool to support packing in columns without fritted disc
- PTFE plug controls flowrate
- Packing not included
Specifications
Specifications
| For Use With (Application) | Analytical separation |
| Diameter (Metric) Inner | 30 mm |
| Type | Plain Chromatography Column |
| Column Capacity | 300 mL |
| Length (Metric) | 500 mm |
| Material | Glass |
| Support Material | Glass Wool |
| Includes | 2mm Bore PTFE Stopcock Plug |