Learn More
DWK Life Sciences Kimble™ Kontes™ Replacement Tube For All-Glass Dounce Tissue Grinders
A range of replacement tubes for Dounce Tissue Grinders, from 2mL to 100mL.
Brand: DWK Life Sciences 885303-0040
This item is not returnable.
View return policy
Description
A range of replacement tubes for Dounce Tissue Grinders, from 2mL to 100mL.
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Case of 1 | |
40 mL | |
Dounce all Glass tissue grinders |
1.35 oz. | |
Tissue Grinder Tube | |
1.35 oz., 40mL |