Learn More
DWK Life Sciences Kimble™ Open-Top PolypropyleneScrew Thread Closures with Red PTFE-Faced Silicone Septa
Autoclavable and used for multiple injections
Brand: DWK Life Sciences 73812BK-9425
This item is not returnable.
View return policy
Description
- Manufactured using the highest quality raw materials
- Designed for an ideal fit with Kimble Chase autosampler vials
- Convenient assembly of caps and liners
- Available with standard or pre-slit septa
Specifications
Black | |
No | |
9-425 |
Polypropylene | |
Autoclavable |