Learn More
DWK Life Sciences Kimble™ Open-Top PolypropyleneScrew Thread Closures with Red PTFE-Faced Silicone Septa
Autoclavable and used for multiple injections
Brand:  DWK Life Sciences 73812WH13425
 View more versions of this product
Product Code. 11833033
  • 856.00 EUR / Pack of 1000
Estimated Shipment: 04-04-2025
to see stock.
Explore more special offers
Description
  • Manufactured using the highest quality raw materials
  • Designed for an ideal fit with Kimble Chase autosampler vials
  • Convenient assembly of caps and liners
  • Available with standard or pre-slit septa
TRUSTED_SUSTAINABILITY
Specifications
Specifications
White
No
13-425
Polypropylene
Autoclavable