Learn More
DWK Life Sciences Kimble™ Open-Top PolypropyleneScrew Thread Closures with Red PTFE-Faced Silicone Septa
Autoclavable and used for multiple injections
Brand:  DWK Life Sciences 73813BK13425
 View more versions of this product
Product Code. 11853033
  • 1040.00 EUR / Pack of 1000
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Description
  • Manufactured using the highest quality raw materials
  • Designed for an ideal fit with Kimble Chase autosampler vials
  • Convenient assembly of caps and liners
  • Available with standard or pre-slit septa
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Black
Yes
Autoclavable
Polypropylene
Pre-Slit
13-425