Learn More
DWK Life Sciences KIMBLE™ EPA Water Analysis Vial, Without Closure
Product Code. p-204124
Click to view available options
Volume (Metric):
20 mL
30 mL
40 mL
60 mL
Color:
Clear
No. per Pack:
144
Unit Size:
Pack of 144
Pack of 432
Product Code. 12703077
Brand: DWK Life Sciences 60958A4
Only null left
Add to basket
Add to basket
These clear and amber screw thread EPA vials for the collection and storage of water samples comply with the guidelines for establishing test procedures for the analysis of pollutants.
  • Clear EPA vials are Converted from Type I, 33 expansion borosilicate glass tubing and amber EPA vials are converted from Type I, 51 expansion borosilicate glass tubing
  • While vials are not pre-cleaned, they are packaged in a plastic shrink wrap module, lehr-clean, as they come from the end of the production lehr and then are placed into a corrugated shelf-pack
  • Open-top closures and PTFE-faced silicone septa or solid-top PFTE-faced white rubber-lined closures are available separately
Compliance
Vials comply with guidelines for establishing test procedures for analyzing pollutants in water (Ref. U.S. EPA Methods 601, 602, 603, and 604).
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 1.1 in.
Color Clear
Diameter (Metric) Outer 28 mm
Material Borosilicate Glass
Volume (Metric) 20 mL
For Use With (Application) For water analysis
Height (English) 2.24 in.
No. per Pack 144
Volume (English) 0.67 oz.
Closure Type Screw Cap
Height (Metric) 57 mm
Certifications/Compliance EPA 601, 602, 603, 604