Learn More
DWK Life Sciences Kimble™ Kontes™ Flow Adapter Replacement Components for CHROMAFLEX™ Columns
Click to view available options
Diameter (Metric) Inner:
2.5 cm
Unit Size:
Pack of 10
Description
- HDPE Bed Support
- 20μm HDPE Bed Support
- 1.0cm, 2.5cm and 4.8cm options
- Case of 10
Specifications
Specifications
| Diameter (Metric) Inner | 2.5 cm |
| Material | HDPE |