Learn More
Kimberly-Clark™ Kimcare Industrie™ Force Solvent Free Hand Cleanser
Click to view available options
Capacity (Metric):
3.5 L
Unit Size:
Pack of 2
Description
- With walnut shells and moisturiser
- Tough on removing dirt
- Gentle on skin
- Advanced formula tough heavy oil, grease, paint and most inks
- For busy working environments
- Cost-effective solution
Specifications
Specifications
| Product Type | Hand Cleanser |
| Color | Pink |
| Dimensions (L x W x H) | 24.4 x 17.23 x 13.2 cm |
| Width (Metric) | 17.23 cm |
| Height (Metric) | 13.2 cm |
| Capacity (Metric) | 3.5 L |
| For Use With (Equipment) | Kimberly-Clark Professional hand cleanser dispenser |
| Quantity | 2 Pk. |