missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIN Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
193.30 EUR - 457.87 EUR
Specifications
Antigen | KIN |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18331506
|
Bio-Techne
NBP3-04797-20UL |
20 μL |
193.30 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18361887
|
Bio-Techne
NBP3-04797-100UL |
100 μL |
457.87 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KIN Polyclonal specifically detects KIN in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
KIN | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
22944 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
antigenic determinant of recA protein (mouse) homolog, Binding to curved DNA, BTCDDNA/RNA-binding protein KIN17, KIN, antigenic determinant of recA protein homolog, KIN, antigenic determinant of recA protein homolog (mouse), KIN17antigenic determinant of recA protein homolog | |
Recombinant fusion protein containing a sequence corresponding to amino acids 180-270 of human KIN (NP_036443.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |