Learn More
Kimberly-Clark Professional™ KleenGuard™ A20+ Coveralls
Provides general protection against powders, larger fibres, radioactive dust and chemical splashes. Kimberly-Clark™ Kleenguard™ A20+ Coveralls are perfect for general maintenance, cleaning and construction utility areas.
179.10 EUR - 185.16 EUR
Specifications
Product Type Single-use Coveralls
Gender Unisex
Disposable Single-use
Sterility Non-sterile
View More Specs
Products 2
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
13474556
View Documents
Kimberly-Clark Professional™
95150
Small
185.16 EUR
Pack of 25
Estimated Shipment: 24-03-2025
to see stock.
Add to basket
13460759
View Documents
Kimberly-Clark Professional™
95200
3X-Large
179.10 EUR
Pack of 25
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
Description
Description
  • Breathable, silicone-free coveralls, with an elasticated hood, ankle and wrists for comfort, safety and strength
  • Approved as Complex Design (Category 3) equipment - offering protection to the levels specified for Type 5 and 6 by CEN
  • Protects against powders, larger fibres, radioactive dust and chemical splashes
  • Suitable for use as protection against solid particles and particulate radioactive contamination - EN1073-2
  • Silicone-free, low lint and treated to be antistatic - to EN 1149-5
  • Single-use protective clothing to improve safety and comfort in high-risk areas. Reduces contamination in critical areas
  • Offers complete freedom of movement with comfortable body design
  • Ideal for working in general manufacturing and maintenance
  • Tripled-stitched seams help to protect against tearing with an easy-fastening snap front
Specifications
Specifications
Single-use Coveralls
Unisex
Single-use
Non-sterile