missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLF12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528010
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2, mouse kidney. Predicted reactivity: Mouse (97%), Rat (90%), Pig (98%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Activator protein-2 alpha (AP-2 alpha) is a developmentally-regulated transcription factor and important regulator of gene expression during vertebrate development and carcinogenesis. The protein encoded by this gene is a member of the Kruppel-like zinc finger protein family and can repress expression of the AP-2 alpha gene by binding to a specific site in the AP-2 alpha gene promoter. Repression by the encoded protein requires binding with a corepressor, CtBP1. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| KLF12 | |
| Polyclonal | |
| Unconjugated | |
| KLF12 | |
| 2700063E05Rik; AI225908; AI315654; AP-2 repressor; AP2REP; AP-2rep; AP-2rep transcription factor; B130052C06Rik; BHLHB24; D530033K05Rik; HSPC122; ID; KLF12; KLF12 zinc finger transcriptional repressor; Krueppel-like factor 12; Kruppel like factor 12; Kruppel-like factor 12; transcriptional repressor AP-2rep | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11278, 16597 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.71 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| O35738, Q9Y4X4 | |
| KLF12 | |
| Recombinant protein encompassing a sequence within the center region of human KLF12. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction