missing translation for 'onlineSavingsMsg'
Learn More

Kobe2602, MedChemExpress

Product Code. 16412847
Encompass_Preferred
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
This item is not returnable. View return policy

Product Code. 16412847

Brand: Medchem Express HY15717

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Kobe2602 is a Ras-Raf interaction inhibitor. Kobe2602 inhibits the binding of H-Ras·GTP to c-Raf-1 RBD with a Ki of 149 μM. Kobe2602 has antitumor activity.

Kobe2602 is a Ras-Raf interaction inhibitor. Kobe2602 inhibits the binding of H-Ras·GTP to c-Raf-1 RBD with a Ki of 149 μM. Kobe2602 has antitumor activity.

Specifications

Chemical Name or Material Kobe2602
CAS 454453-49-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C14H9F4N5O4S
Quantity 50 mg
Solubility Information DMSO : 14.3 mg/mL (34.10 mM; Need ultrasonic and warming)
SMILES FC(C1=CC([N+]([O-])=O)=C(NNC(NC2=CC=C(F)C=C2)=S)C([N+]([O-])=O)=C1)(F)F
Molecular Weight (g/mol) 419.31
Formula Weight 419.31
Percent Purity 99.33%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.