Learn More
Kontes™ Membrane Filtration Glassware from Restek
Product Code. 11739055
Click to view available options
Type:
Flask (2 L)
Flask (4 L)
Funnel (1 L)
Funnel (300 mL)
Funnel (500 mL)
Support
Material:
Fritted Glass
Glass
Unit Size:
Each
Product Code. 11739055
Brand: Kimble Chase 9538260000
Used for routine filtration of corrosive liquids and removal of particles from HPLC solvents
  • Ground joint connection eliminates phthalate contamination that can occur when using silicone or neoprene stoppers
  • Support base has a coarse porosity glass frit and an integral vacuum connection, located above the drip tip that prevents contamination of the vacuum line with filtrate droplets
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Microfiltration Apparatus
Material Fritted Glass
Standard Taper Joint 40/35
Type Support
Length (Metric) 47 mm