Learn More
DWK Life Sciences Kimble™ Kontes™ NMR Polyethylene Sample Tube Pressure Caps for 5 and 10mm Tubes
Tight-fitting, polyethylene cap which stays in place even with a build-up of internal vaporization pressures
Brand:  DWK Life Sciences 897095-0001
 View more versions of this product
Product Code. 11873143
  • 28.95 EUR / Pack of 100
Estimated Shipment: 10-06-2025
to see stock.
Explore more special offers
Specifications
Pressure Cap
Red
5 mm NMR Sample Tube
Polyethylene
Provides tight-fitting and stays in place even with a build-up of internal vaporization pressures