Learn More
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (90mm)
Designed for large volume filtration analysis of corrosive liquids or samples with high particulate loads that would tend to clog a 47 mm diameter filter
Brand:  DWK Life Sciences 953825-0090
 View more versions of this product
Product Code. 11867513
  • 2485.00 EUR / Each
Estimated Shipment: 10-09-2025
to see stock.
Explore more special offers
Description
This unit is also recommended for filtering large volumes of HPLC solvents.
  • The ground glass connection eliminates the possibility of phthalate contamination that can occur when using silicone or neoprene stoppers
      Each apparatus is supplied with 1000mL funnel; 90mm, 40-60 micron porosity, fritted glass support base; anodized aluminum clamp; and filtration flask
    • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
    TRUSTED_SUSTAINABILITY
Specifications
Specifications
Microfiltration Assembly
Glass
1 L
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products