missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KV1.5 (KCNA5) (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577650
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
In neurons voltage-dependent potassium channels are key determits of the resting membrane potential and of membrane excitability, conditioning the frequency, and shape of action potential.
Specifications
| KV1.5 (KCNA5) (extracellular) | |
| Polyclonal | |
| Unconjugated | |
| KCNA5 | |
| KCNA5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25470, 3741 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| P19024, P22460 | |
| KCNA5 | |
| Peptide (C)DERELLRHPPVP(K), corresponding to amino acid residues 268-279 of rat Kv 1.5 | |
| 50 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction