missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv6.4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | Kv6.4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Kv6.4 Polyclonal specifically detects Kv6.4 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Kv6.4 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
93107 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
KV6.3, KV6.4, MGC129609, potassium voltage-gated channel, subfamily G, member 4, voltage-gated potassium channel Kv6.3, voltage-gated potassium channel subunit Kv6.4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human Kv6.4 (NP_758857.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |