missing translation for 'onlineSavingsMsg'
Learn More

KY1220, MedChemExpress

Product Code. p-200098009
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16494766

Brand: Medchem Express HY102028

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KY1220 is a compound that destabilizes both β-catenin and Ras, via targeting the Wnt/β-catenin pathway; with an IC50 of 2.1 μM in HEK293 reporter cells.

KY1220 is a compound that destabilizes both β-catenin and Ras, via targeting the Wnt/β-catenin pathway

Specifications

Chemical Name or Material KY1220
CAS 292168-79-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C14H10N4O3S
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (318.15 mM)
SMILES O=C(/C(N1)=C/C2=CC=CN2C3=CC=C([N+]([O-])=O)C=C3)NC1=S
Molecular Weight (g/mol) 314.32
Formula Weight 314.32
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.