Learn More
L-Arginine methyl ester dihydrochloride, 98%, Thermo Scientific Chemicals
48.14 EUR - 155.78 EUR
Chemical Identifiers
CAS | 26340-89-6 |
---|---|
Molecular Formula | C7H18Cl2N4O2 |
Molecular Weight (g/mol) | 261.147 |
MDL Number | MFCD00038948 |
InChI Key | XQYZOBNLCUAXLF-XRIGFGBMSA-N |
PubChem CID | 117776 |
IUPAC Name | methyl (2S)-2-amino-5-(diaminomethylideneamino)pentanoate;dihydrochloride |
SMILES | COC(=O)C(CCCN=C(N)N)N.Cl.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11352306
|
Thermo Scientific Alfa Aesar
L05959.06 |
5 g |
48.14 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11362306
|
Thermo Scientific Alfa Aesar
L05959.14 |
25 g |
155.78 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
26340-89-6 | |
261.147 | |
XQYZOBNLCUAXLF-XRIGFGBMSA-N | |
methyl (2S)-2-amino-5-(diaminomethylideneamino)pentanoate;dihydrochloride |
C7H18Cl2N4O2 | |
MFCD00038948 | |
117776 | |
COC(=O)C(CCCN=C(N)N)N.Cl.Cl |
Specifications
∽190°C (decomposition) | |
C7H18Cl2N4O2 | |
4159929 | |
XQYZOBNLCUAXLF-XRIGFGBMSA-N | |
methyl (2S)-2-amino-5-(diaminomethylideneamino)pentanoate;dihydrochloride | |
117776 | |
+21° (c=2.5 in Methanol) | |
98% |
26340-89-6 | |
MFCD00038948 | |
l-arginine methyl ester dihydrochloride, h-arg-ome.2hcl, methyl l-argininate dihydrochloride, h-arg-ome 2hcl, l-arginine, methyl ester, dihydrochloride, s-methyl 2-amino-5-guanidinopentanoate dihydrochloride, methyl 2s-2-amino-5-carbamimidamidopentanoate dihydrochloride, l-arginine, methyl ester, hydrochloride 1:2, h-arg-ome?hcl, h-arg-ome 2 hcl | |
COC(=O)C(CCCN=C(N)N)N.Cl.Cl | |
261.147 | |
261.15 | |
Hygroscopic | |
L-Arginine methyl ester dihydrochloride |
Safety and Handling
missing translation for 'einecsNumber' : 247-622-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.