missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic Acid Sodium Salt, MP Biomedicals

Product Code. 11436272 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
500 g
1 kg
Unit Size:
1kg
25g
500g
This item is not returnable. View return policy

Product Code. 11436272

Brand: MP Biomedicals 0210289090

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 134-03-2
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
MDL Number MFCD00082340
InChI Key IFVCRSPJFHGFCG-HXPAKLQESA-N
Synonym sodium ascorbate, l-ascorbic acid sodium salt, sodium l-ascorbate, vitamin c sodium, ascorbicin, sodascorbate, cebitate, aminofenitrooxon, natrii ascorbas, monosodium l-ascorbate
PubChem CID 131674100
IUPAC Name sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
SMILES [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

Specifications

Color White
Assay Percent Range ≥99%
Quantity 500 g
Formula Weight 198.11
Physical Form Powder
Chemical Name or Material L-Ascorbic Acid Sodium Salt, Antimicrobial, Antioxidant, Reductant for the isolation of chloroplasts
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.