missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine, 99-100%, MP Biomedicals

Product Code. 11425202
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
PubChem CID 5961
ChEBI CHEBI:18050
IUPAC Name (2S)-2-amino-4-carbamoylbutanoic acid
SMILES N[C@@H](CCC(N)=O)C(O)=O

Specifications

Grade Cell Culture Reagent
Quantity 1 kg
Physical Form Crystalline
Chemical Name or Material L-Glutamine, 99-100%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'