Learn More
Thermo Scientific Chemicals L-Lysine, 98%
91.20 EUR - 691.00 EUR
Chemical Identifiers
CAS | 56-87-1 |
---|---|
Molecular Formula | C6H14N2O2 |
Molecular Weight (g/mol) | 146.19 |
MDL Number | MFCD00064433 |
InChI Key | KDXKERNSBIXSRK-MLHKIVSYNA-N |
Synonym | l-lysine, lysine, h-lys-oh, lysine acid, s-lysine, aminutrin, 2s-2,6-diaminohexanoic acid, l-+-lysine, alpha-lysine, lysinum |
PubChem CID | 5962 |
ChEBI | CHEBI:18019 |
SMILES | NCCCC[C@H](N)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15404639
|
Thermo Scientific Chemicals
|
25 g |
91.20 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15414639
|
Thermo Scientific Chemicals
J62225.22 |
100 g |
263.00 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15424639
|
Thermo Scientific Chemicals
J62225.36 |
500 g |
691.00 EUR
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
L-Lysine is an essential amino acid used in human nutrition. It plays a vital role in calcium absorption, building muscle protein and recovering from surgery or sports injuries. It is used for the treatment of herpes infections and cold sores. Its derivatives lysine acetylsalicylate is utilized to treat pain as well as to detoxify the body after heroin use. It is an important additive to animal feed. Further, it is used in many foods, especially red meats, fish and dairy products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Lysine is an essential amino acid used in human nutrition. It plays a vital role in calcium absorption, building muscle protein and recovering from surgery or sports injuries. It is used for the treatment of herpes infections and cold sores. Its derivatives lysine acetylsalicylate is utilized to treat pain as well as to detoxify the body after heroin use. It is an important additive to animal feed. Further, it is used in many foods, especially red meats, fish and dairy products.
Solubility
Soluble in water. Insoluble in ethanol, ethyl ether, acetone, benzene and common neutral solvent.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Chemical Identifiers
56-87-1 | |
146.19 | |
KDXKERNSBIXSRK-MLHKIVSYNA-N | |
5962 | |
NCCCC[C@H](N)C(O)=O |
C6H14N2O2 | |
MFCD00064433 | |
l-lysine, lysine, h-lys-oh, lysine acid, s-lysine, aminutrin, 2s-2,6-diaminohexanoic acid, l-+-lysine, alpha-lysine, lysinum | |
CHEBI:18019 |
Specifications
215°C | |
White | |
MFCD00064433 | |
14,5636 | |
Soluble in water; Insoluble in ethanol,ethyl ether,acetone,benzene and common neutral solvent. | |
NCCCC[C@H](N)C(O)=O | |
146.19 | |
CHEBI:18019 | |
98% | |
Powder |
56-87-1 | |
C6H14N2O2 | |
1722531 | |
l-lysine, lysine, h-lys-oh, lysine acid, s-lysine, aminutrin, 2s-2,6-diaminohexanoic acid, l-+-lysine, alpha-lysine, lysinum | |
KDXKERNSBIXSRK-MLHKIVSYNA-N | |
(2S)-2,6-diaminohexanoic acid | |
5962 | |
146.19 | |
+26° (c=2 in 6M HCl) | |
L-Lysine |
Safety and Handling
missing translation for 'einecsNumber' : 200-294-2
missing translation for 'rtecsNumber' : OL5540000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only