missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-lysine, free base, MP Biomedicals™
Click to view available options
Quantity:
100 g
Specifications
Specifications
CAS | 39665-12-8 |
Color | White |
Molecular Formula | C6H16N2O3 |
MDL Number | MFCD00151035 |
Synonym | l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1 |
InChI Key | HZRUTVAFDWTKGD-UHFFFAOYNA-N |
SMILES | O.NCCCCC(N)C(O)=O |
IUPAC Name | 2,6-diaminohexanoic acid hydrate |
Molecular Weight (g/mol) | 164.21 |
PubChem CID | 16211825 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction