missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-(-)-Mannose, 99%

Catalog Number p-7023809
Click to view available options
Quantity:
0.25 g
1 g
This item is not returnable. View return policy
10030-80-5
C6H12O6
180.16
MFCD00136021
GZCGUPFRVQAUEE-BXKVDMCESA-N
l-mannose, l---mannose, aldehydo-l-mannose, mannose, l, 2r,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, unii-2w3ye50tx8, aldehydo-l-manno-hexose, l-?-mannose
82308
CHEBI:37681
(2R,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal
OC[C@H](O)[C@H](O)[C@@H](O)[C@@H](O)C=O
This item is not returnable. View return policy
10030-80-5
C6H12O6
180.16
MFCD00136021
GZCGUPFRVQAUEE-BXKVDMCESA-N
l-mannose, l---mannose, aldehydo-l-mannose, mannose, l, 2r,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, unii-2w3ye50tx8, aldehydo-l-manno-hexose, l-?-mannose
82308
CHEBI:37681
(2R,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal
OC[C@H](O)[C@H](O)[C@@H](O)[C@@H](O)C=O

L-Mannose is used in a study to assess substrate specificity of galactokinase from Streptococcus pneumoniae. It has also been used in a study to investigate a versatile route to L-hexoses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Mannose is used in a study to assess substrate specificity of galactokinase from Streptococcus pneumoniae. It has also been used in a study to investigate a versatile route to L-hexoses.

Solubility
Soluble in Water (100 mg/mL).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point 129°C to 131°C
Quantity 0.25 g
Beilstein 1724628
Solubility Information Soluble in Water (100mg/mL).
Formula Weight 180.16
Percent Purity 99%
Chemical Name or Material L-(-)-Mannose

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'