missing translation for 'onlineSavingsMsg'
Learn More

Melford L-Ornithine HCL

Product Code. p-200087138
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 30018638

Brand: Melford O20040100.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Polyamine growth regulator.

Polyamine growth regulator.

Specifications

CAS 3184-13-2
Molecular Formula C5H12N2O2-HCl
Quantity 100 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.