Learn More
Masterflex™ L/S™ 2-Stop Precision Pump Tubing
Masterflex™ L/S™ 2-Stop Precision Pump Tubing with excellent biocompatibility that meets multiple classifications
Brand:  Masterflex™ 06421-25
Product Code. 11866743
  • 188.00 EUR / Pack of 8
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Description
  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance and pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match all unique applications
  • Always in stock - most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Slightly greater clarity than peroxide-cured tubing
  • Smooth surfaces; lower protein binding levels
  • Fewer potential leachables
  • Tubing Size: L/S 25
Specifications
Specifications
Platinum-cured Silicone
≤ 1000 mL/min
8/Pk.
L/S 25
3/16 in.
660 mm Hg