Learn More
Masterflex™ L/S™ Bulk-Packed Platinum-Cured Silicone High-Performance Precision Pump Tubing
Excellent biocompatibility that meets multiple classifications
Brand:  Masterflex™ 96404-24
 View more versions of this product
Product Code. 11750359
  • 2870.00 EUR / Pack of 20
Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers
Description
Description
  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Slightly greater clarity than peroxide-cured tubing
  • Smooth surfaces; lower protein binding levels
  • Fewer potential leachables
  • Pharmaceutical, biotech, peristaltic pumping, and food applications
  • Excellent heat resistance, low compression set, odorless, nontoxic, translucent
  • Comes in convenient cases containing up to 20 sealed bags of tubing (depending on tubing size).
  • Bulk tubing eliminates individual cardboard boxes, reduces unpacking time, and saves you money on shipping.
  • Tubing comes in 25-ft bags.
Specifications
Specifications
Platinum-cured Silicone
7.6 m
-40°C
1800 mL/min
100°C
25 ft.
Pharmaceutical, Biotech, Peristaltic Pumping, and Food Applications
24
1/4 in.