missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ L/S™ Pump Head Adapter

Masterflex™ L/S™ Pump Head Adapter for mounting L/S™ Pump Heads to a 56C Frame Motor

Brand:  Masterflex™ 77495-20

Product Code. 11865191

  • 1180.00 EUR / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Customize your pump by mounting L/S™ Pump Heads to a 56C Frame Motor (pump head and motor sold separately)
  • Use up to 2 L/S™ Pump Heads to pump two fluids simultaneously at equal or different flow rates
  • IP56-rated cast-aluminum adapters feature built-in gear box
  • Flow range 0.06 to 1100 mL/min - flow rate depends on drive rpm, tubing head, and tubing size
  • Adapters accept seven different pump head types: Standard, Easy-Load™, Easy-Load™ II, High-Performance, Cartridge, PTFE-Tubing, and PTFE-Diaphragm
Specifications

Specifications

18.3:1 Gear Ratio
0.06 to 1100 mL/min.
Painted Aluminum
1/4 hp min, 3450 rpm max
Direct Coupling Mounting
(146 x 165 x 165 mm)
Masterflex™ L/S™ Pump Heads
NEMA Type 56C
2 Pump Heads Accepted
1-90 rpm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.