missing translation for 'onlineSavingsMsg'
Learn More

L-Tryptophan, ≥99%, MP Biomedicals™

Product Code. 11413581 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
5g
This item is not returnable. View return policy

Product Code. 11413581

Brand: MP Biomedicals 0210315105

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Tryptophan is an amino acid precursor of serotonin and melatonin

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-22-3
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD00064340
InChI Key QIVBCDIJIAJPQS-VIFPVBQESA-N
Synonym l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
PubChem CID 6305
ChEBI CHEBI:16828
IUPAC Name (2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Color White
Assay Percent Range ≥99%
Quantity 5 g
Formula Weight 204.22
Physical Form Crystalline Powder
Chemical Name or Material L-Tryptophan
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.