missing translation for 'onlineSavingsMsg'
Learn More

Androgen Receptor Ab-1 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation, and gel supershift experiments.

Brand:  Epredia MS-443-R7

 View more versions of this product

Product Code. 12661437

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

The expression of AR is reportedly inversely correlated with histologic grade, i.e. well differentiated prostate tumors show higher expression than the poorly differentiated tumors. In prostate cancer, AR has been proposed as a marker of hormone-responsiveness.

Host Species: Mouse

Clone: AR 441

Isotype: IgG1

Species Reactivity: Human and Dog. Does not react with mouse. Others not tested.

Epitope: aa 299-315

Immunogen: A synthetic peptide from human AR

Molecular Weight: 110kDa

Positive Control: LnCAp cells or prostate CA

Cellular Localization: Nuclear

Ab-1 is excellent for staining of formalin/paraffin tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Androgen Receptor Ab-1
Monoclonal
Unconjugated
A synthetic peptide from human AR
RUO
Primary
Prediluted
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
AR 441
Mouse
7 mL
Steroids Hormones and Growth Factors
Canine, Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.