missing translation for 'onlineSavingsMsg'
Learn More

MGMT (O-methylguanine-DNA methyltransferase) Ab-1 Mouse Monoclonal Antibody, Epredia™

Epredia™ MGMT (O-methylguanine-DNA methyltransferase) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-470-PABX

 View more versions of this product

Product Code. 12621537

This item is not returnable. View return policy

Description

Description

Cancer chemotherapeutic alkylating agents (e.g. BCNU,) act by inducing formation of lethal cross-links at the O-alkylguanine position in DNA. MGMT transfers alkyl adducts from the O-position of guanine in DNA (prior to cross-link formation) to a cysteine residue in its own sequence, thereby restoring DNA to its intact state. This transfer inactivates the MGMT enzyme and is irreversible; hence the level of MGMT in a cell is directly proportional to the level of DNA-damage it can tolerate. In normal tissues, MGMT acts as a suppressor of mutation and carcinogenesis. Tumors with high levels of MGMT are likely be drug-resistant.

Host Species: Mouse

Clone: MT 3.1

Isotype: IgG1

Species Reactivity: Human. Also reacts weakly with Mouse, Rat and Hamster. Others not known.

Epitope: aa 8-221

Immunogen: Purified recombinant human MGMT protein

Molecular Weight: 25kDa

Positive Control: Raji cells, Colon CA

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

MGMT (O-methylguanine-DNA methyltransferase) Ab-1
Monoclonal
1mg/mL
Mouse
200 μL
Enzyme
Hamster, Human, Mouse, Rat
IgG1
Immunohistochemistry (Paraffin), Western Blot
MT 3.1
Unconjugated
Purified recombinant human MGMT protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.