Learn More
Epredia™ Lab Vision™ PT Module™ Deparaffinization and Heat-Induced Epitope Retrieval Solutions (100X), Citrate buffer, pH 6
Shop All Epredia Products
Click to view available options
Quantity:
125mL
250mL
pH:
6
8
9
Unit Size:
125mL
250mL
Description
- 100X stock solution
- Dilute 100-fold with distilled water before use
- Designed for use with Pretreatment Module™
- Available in Citrate, EDTA or Tris-HCl at various pH levels
Storage Conditions: Store at room temperature. Stable for up to 18 months. Contains no preservatives; for periods longer than three months, store at 2°-8°C.
Specifications
Specifications
| Content And Storage | Room temperature |
| Dilution | 100X |
| Product Type | HIER |
| Quantity | 125mL |
| For Use With (Application) | Immunohistochemistry |
| pH | 8 |