Learn More
Labconco™ Exterior Glove Port Covers
May be placed over glove ports with or without gloves attached to facilitate purge/fill operations and preserve inert atmosphere
Brand: Labconco™ 5242200
Includes: Two clamps
Description
May be placed over glove ports with or without gloves attached to facilitate purge/fill operations and preserve inert atmosphere
- Molded neoprene construction
- For use with all glove boxes
Specifications
3.5 lb. | |
1.6 Kg | |
Enclosure Kit | |
All Glove Box |