Learn More
Labconco™ Freeze-Drying Ampule Accessories
Freeze drying ampule accessories allow ampules to be used with Labconco FreeZone Freeze Dryers.
Brand: Labconco™ 7576210
This item is not returnable.
View return policy
Description
Freeze drying ampule accessories allow ampules to be used with Labconco FreeZone Freeze Dryers.
Large rubber split stoppers fit 5, 10, 20, 30, 50, 100 and 125 ml Labconco Serum Bottles (bottles with 20mm corkage). Package of 100.Specifications
Stopper | |
Rubber | |
Freeze drying system | |
2 cm |