missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lambda Light Chain Mouse, Clone: LLC/1738, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3535-MSM8-P1
This item is not returnable.
View return policy
Specifications
Lambda Light Chain | |
Cell Surface, cytoplasmic and secreted | |
LLC/1738 | |
Unconjugated | |
IGL | |
Purified human full-length lambda light chain | |
293T, Raji or hPBL cells. Tonsil or Spleen | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
Mouse | |
22.5kDa | |
Protein A/G Purified | |
RUO | |
3546 | |
IgG1 κ |