missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin alpha-1 Monoclonal Antibody (4C7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA121194
This item is not returnable.
View return policy
Description
This antibody reacts with the terminal globular domain of the A chain. It does not cross react with laminin from mouse or rat. The staining with this antibody is sensitive to fixation of paraffin tissues and use of proteolytic enzymatic treatment. Recommended positive control: skin tissue.
Laminin antibodies are widely used to label blood vessels and basement membranes.Specifications
Laminin alpha-1 | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 0.09% sodium azide | |
P25391 | |
LAMA1 | |
The A chain of human laminin. | |
RUO | |
284217 | |
4° C, do not freeze | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
4C7 | |
Unconjugated | |
LAMA1 | |
LAMA; LAMA1; Laminin A chain; laminin subunit alpha 1; laminin subunit alpha-1; laminin, alpha 1; Laminin-1 subunit alpha; Laminin-3 subunit alpha; PTBHS; S-LAM alpha; S-LAM-alpha; S-laminin subunit alpha | |
Mouse | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |