Learn More
OHAUS™ Large Three Prong Sleeves
Product Code. p-7158983 Shop All Ohaus Products
Click to view available options
Material:
Fiberglass
Vinyl
Color:
Black
Gray
Red
Unit Size:
Each
Product Code. 15702679
Brand: OHAUS™ 30400261
Only null left
Add to basket
Add to basket
OHAUS™ Large Three Prong Sleeves are for use with clamps and can be easily removed for cleaning or replacement.
  • Available in red or black vinyl and fiberglass
  • Fiberglass sleeves are ideal for application above 100°C
TRUSTED_SUSTAINABILITY
Specifications
Quantity 3
Type Prong Sleeve
Material Vinyl
Size Large
Color Red