Learn More
Fisherbrand™ Latex Dropper Teats
Shop All Fisherbrand Products
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
1 mL
1.5 mL
1.5 to 5 mL (Assorted)
2.5 mL
3.75 mL
missing translation for 'unitSize'
Pack of 20
Specifications
Specifications
| Material | Latex |
| Type | Bulb Dropper |
| Volume (Metric) | 3.75 mL |
| Diameter (Metric) Opening | 6 mm |