Learn More
Lonza™ Latitude™ HT Precast Gels
Ready-to-use gels for high-throughput DNA electrophoresis. Latitude™ HT Precast Gels are precision cast for accurate, reproducible resolution of DNA fragments for 8bp to 10kb with our SeaKem™ or NuSieve™ Agarose.
353.00 EUR
Specifications
Buffer | TBE |
---|---|
Concentration | 4% |
Product Type | Precast Gel |
Separation Range | 8 to ≥1000 bp |
Dnase Free | Yes |
Alert:
Ethidium bromide is a powerful mutagen; gloves should be worn when opening pouches and handling gels and any solutions containing ethidium bromide
Description
- Provide rapid, reproducible DNA resolution of 8 bp to ≥10kb
- Manufactured from high-quality molecular biology grade SeaKem™ and NuSieve™ Brand Agarose
- Quality and performance tested
- Fit most large submerged electrophoresis chambers
- Compatible with 8- and 12-tip multichannel pipets and microtiter plate spacing
- Precision cast in accordance with ISO 9001 standards
- Come in 100- and 200-well formats
- Prestained with 0.5μg/mL of ethidium bromide
- Permit runs to be completed in 30 minutes
- Allow up to 200 samples to be run on a single precast gel
Specifications
TBE | |
Precast Gel | |
Yes | |
High throughput DNA electrophoresis | |
5 Gels | |
18° to 26°C | |
0.65 cm | |
24 cm |
4% | |
8 to ≥1000 bp | |
4% NuSieve 3:1 Plus Agarose, 1X TBE, Prestained with 0.5μg/mL of Ethidium Bromide | |
Agarose | |
Yes | |
0.25 in. | |
10 to 12 μL | |
14 cm |
Safety and Handling
missing translation for 'shelfLife' : 6 to 12 Months
Your input is important to us. Please complete this form to provide feedback related to the content on this product.