missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lauda™ Alpha 2 Immersion Circulators
Brand: Lauda L000618
840.83 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Specifications
CE | |
Pressure | |
LED | |
0.2 bar | |
100°C | |
125 mm | |
150 mm | |
230 V 50/60 Hz | |
50/60 Hz |
15 L/min. | |
25°C to 100°C | |
Sample preparation for chemico-pharmaceutical analysis, precise temperature control in sensitive are | |
LCE0226 | |
±0.05K | |
300 mm | |
3.5 kg | |
230 V |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction