missing translation for 'onlineSavingsMsg'
Learn More

Lauric acid, sodium salt, 98%, Thermo Scientific Chemicals

Product Code. 10250031
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-25-4
Molecular Formula C12H23NaO2
Molecular Weight (g/mol) 222.30
MDL Number MFCD00041754
InChI Key BTURAGWYSMTVOW-UHFFFAOYSA-M
Synonym sodium dodecanoate, sodium laurate, lauric acid sodium salt, dodecanoic acid, sodium salt, lauric acid, sodium salt, dodecanoic acid sodium salt, lauran sodny czech, caswell no. 778a, unii-k146mr5exo, epa pesticide chemical code 079026
PubChem CID 2735067
IUPAC Name sodium;dodecanoate
SMILES [Na+].CCCCCCCCCCCC([O-])=O

Specifications

Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)10COONa
Quantity 25 g
Formula Weight 222.3
Percent Purity 98%
Physical Form Powder or Flakes
Chemical Name or Material Lauric acid, sodium salt, 99%
compliance-icons compliance-icons
Product Identifier
  • Lauric acid, sodium salt
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.