missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiCl Saturated in Glacial Acetic Acid Electrolyte Solution, SI Analytics™
Click to view available options
Quantity:
250 mL
Description
- Hazard pictograms: GHS02, GHS05, GHS07
- GHS02 flame: H226 Flammable liquid and vapour
- GHS05 corrosion: H314 Causes severe skin burns and eye damage, H318 Causes serious eye damage
- GHS07: H312 Harmful in contact with skin, H332 Harmful if inhaled
- Precautionary statements: P280 Wear protective gloves/protective clothing/eye protection/face protection. P303+P361+P353 IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower. P305+P351+P338 IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. P309+P311 IF exposed or if you feel unwell: Call a POISON CENTER or doctor/physician. P301+P330+P331 IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- Vapors can form explosive mixture with air above 40°C
- Absorb with liquid-binding material (sand, diatomite, acid binders, universal binders, sawdust)
Specifications
Specifications
CAS | CAS: 64-19-7, CAS: 7447-41-8 |
Melting Point | 16°C |
Boiling Point | 119°C |
Color | Colorless |
Density | 1.09g/cm3 |
Concentration | 0.1 M |
Quantity | 250 mL |
Solution Type | Electrolyte |
Odor | Pungent |
Vapor Pressure | 16hPa |
Show More |
Safety and Handling
Product Identifier
- acetic acid saturated with lithium chloride
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- H226-Flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- MIXTURE LIST-Contains : acetic acid, lithiumchloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction